Learn More
ATCC Human herpesvirus 2; HSV-2 Strain: MS; Quantitative DNA
Catalog No. 502384265
Encompass
Quantitative genomic DNA from Human herpesvirus 2 strain MS can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4265 Supplier ATCC Supplier No. VR540DQ
Quantitative genomic DNA from Human herpesvirus 2 strain MS can be used for assay development, verification, and validation as well as monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.