Learn More
ATCC Human herpesvirus 2; Strain: ATCC-2011-2; Herpes simplex virus 2
Catalog No. 502384395
Encompass
Human herpesvirus 2 strain ATCC-2011-2 is propagated in Vero cells (ATCC CCL-81). The virus was isolated from a clinical sample from a patient positive for HSV-2 in Virginia in 2011. It has applications in infectious disease and sexually transmitted disease research.
Catalog No. 50-238-4395 Supplier ATCC Supplier No. VR1779
Human herpesvirus 2 strain ATCC-2011-2 is propagated in Vero cells (ATCC CCL-81). The virus was isolated from a clinical sample from a patient positive for HSV-2 in Virginia in 2011. It has applications in infectious disease and sexually transmitted disease research.