Learn More
ATCC Human herpesvirus 2; Strain: MS
Catalog No. 502384159
Encompass
Human herpesvirus 2 strain MS, or Herpes simplex virus 2 (HSV-2), is propagated in Vero cells (ATCC CCL-81). This strain was isolated in 1961 from the brain of a 50-year-old, female patient with multiple sclerosis in Iceland and was deposited by the Centers for Disease Control and Prevention. This product has applications in infectious disease research and sexually transmitted disease research.
Catalog No. 50-238-4159 Supplier ATCC Supplier No. VR540
Human herpesvirus 2 strain MS, or Herpes simplex virus 2 (HSV-2), is propagated in Vero cells (ATCC CCL-81). This strain was isolated in 1961 from the brain of a 50-year-old, female patient with multiple sclerosis in Iceland and was deposited by the Centers for Disease Control and Prevention. This product has applications in infectious disease research and sexually transmitted disease research.