Learn More
ATCC Human herpesvirus 3 DNA; VZV strain Ellen
Supplier:  ATCC VR1367D
Encompass
Viral nucleic acids, including virus genomic DNA, in total DNA isolated from MRC-5 cells (ATCC® CCL-171™) infected with Human herpesvirus 5 (VZV) strain Ellen (ATCC® VR-1367™). Virus and host cells are also available through the ATCC® Catalog. This product was prepared using methods known to inactivate viruses.
Catalog No. 50-238-3676