Learn More
ATCC Human herpesvirus 3; Varicella zoster virus; Strain: Ellen
Catalog No. 502383011
Encompass
Human herpesvirus 3 strain Ellen is a mycoplasma-free strain derived from ATCC VR-586 by three passages in the presence of a mycoplasma removal agent. The strain was originally isolated from the vesicular fluid from a child with the chickenpox in Georgia, United States, and can be propagated in MRC-5 cells (ATCC CCL-171). Human herpesvirus 3 has applications in infectious disease research.
Catalog No. 50-238-3011 Supplier ATCC Supplier No. VR1367
Human herpesvirus 3 strain Ellen is a mycoplasma-free strain derived from ATCC VR-586 by three passages in the presence of a mycoplasma removal agent. The strain was originally isolated from the vesicular fluid from a child with the chickenpox in Georgia, United States, and can be propagated in MRC-5 cells (ATCC CCL-171). Human herpesvirus 3 has applications in infectious disease research.