Learn More
ATCC Human herpesvirus 3; VZV Strain Ellen; Quantitative DNA
Catalog No. 502383678
Encompass
Quantitative Genomic DNA from Human herpesvirus 3 (HHV-3) strain Ellen can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3678 Supplier ATCC Supplier No. VR1367DQ
Add to Cart
Add to Cart
Quantitative Genomic DNA from Human herpesvirus 3 (HHV-3) strain Ellen can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.