Learn More
ATCC Human herpesvirus 3;Strain: Oka;Varicella zoster virus
Catalog No. 502382271
Encompass
Human herpesvirus 3 strain Oka is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from a vesicle from a 3-year-old male in Japan and was deposited by the Research Foundation for Microbial Diseases of Osaka University. It has applications in infectious disease research, assay development, and vaccine development.
Catalog No. 50-238-2271 Supplier ATCC Supplier No. VR1832
Human herpesvirus 3 strain Oka is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from a vesicle from a 3-year-old male in Japan and was deposited by the Research Foundation for Microbial Diseases of Osaka University. It has applications in infectious disease research, assay development, and vaccine development.