Learn More
ATCC Human herpesvirus 5; HCMV strain AD-169; DNA
Supplier:  ATCC VR538D
Encompass
Viral nucleic acids, including virus genomic DNA, in total DNA isolated from MRC-5 cells (ATCC CCL-171™?) infected with Human herpesvirus 5 (HCMV) strain AD-169 (ATCC VR-538™?).Virus and host cells are also available through the ATCC Catalog.  This product was prepared using methods known to inactivate viruses.
Catalog No. 50-238-3442