Learn More
ATCC Human herpesvirus 5; Strain: Davis; Human cytomegalovirus
Catalog No. 502384589
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Human herpesvirus 5 strain Davis is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the liver biopsy of a 3-month-old human female with microencephaly and other symptoms resembling congenital toxoplasmosis. This product has applications in infectious disease research.
Catalog No. 50-238-4589 Supplier ATCC Supplier No. VR807
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Human herpesvirus 5 strain Davis is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the liver biopsy of a 3-month-old human female with microencephaly and other symptoms resembling congenital toxoplasmosis. This product has applications in infectious disease research.