Learn More
ATCC Human herpesvirus 6B; Human herpesvirus 6B; Strain: Z29
Supplier:  ATCC 502385112
Encompass
Human herpesvirus 6B strain Z-29 is propagated in MOLT-3 cells (ATCC CRL-1552). This strain was isolated from the peripheral blood lymphocytes of a 36-year-old, male AIDS patient in Zaire. It has applications in infectious disease research.
Catalog No. 50-238-5112