Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HERPUD1 Full-length ORF (AAH00086, 1 a.a. - 391 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-298 10 μg
89-947-299 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-298 Supplier Abnova™ Supplier No. H00009709P02S
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The accumulation of unfolded proteins in the endoplasmic reticulum (ER) triggers the ER stress response. This response includes the inhibition of translation to prevent further accumulation of unfolded proteins, the increased expression of proteins involved in polypeptide folding, known as the unfolded protein response (UPR), and the destruction of misfolded proteins by the ER-associated protein degradation (ERAD) system. This gene may play a role in both UPR and ERAD. Its expression is induced by UPR and it has an ER stress response element in its promoter region while the encoded protein has an N-terminal ubiquitin-like domain which may interact with the ERAD system. This protein has been shown to interact with presenilin proteins and to increase the level of amyloid-beta protein following its overexpression. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined. [provided by RefSeq]

Sequence: MESETEPEPVTLLVKSPNQRHRDLELSGDRGWSVGHLKAHLSRVYPERPRPEDQRLIYSGKLLLDHQCLRDLLPKQEKRHVLHLVCNVKSPSKMPEINAKVAESTEEPAGSNRGQYPEDSSSDGLRQREVLRNLSSPGWENISRPEAAQQAFQGLGPGFSGYTPYGWLQLSWFQQIYARQYYMQYLAATAASGAFVPPPSAQEIPVVSAPAPAPIHNQFPAENQPANQNAAPQVVVNPGANQNLRMNAQGGPIVEEDDEINRDWLDWTYSAATFSVFLSILYFYSSLSRFLMVMGATVVMYLHHVGWFPFRPRPVQNFPNDGPPPDVVNQDPNNNLQEGTDPETEDPNHLPPDRDVLDGEQTSPSFMSTAWLVFKTFFASLLPEGPPAIAN

Specifications

Accession Number AAH00086
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9709
Molecular Weight (g/mol) 68.75kDa
Name HERPUD1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MESETEPEPVTLLVKSPNQRHRDLELSGDRGWSVGHLKAHLSRVYPERPRPEDQRLIYSGKLLLDHQCLRDLLPKQEKRHVLHLVCNVKSPSKMPEINAKVAESTEEPAGSNRGQYPEDSSSDGLRQREVLRNLSSPGWENISRPEAAQQAFQGLGPGFSGYTPYGWLQLSWFQQIYARQYYMQYLAATAASGAFVPPPSAQEIPVVSAPAPAPIHNQFPAENQPANQNAAPQVVVNPGANQNLRMNAQGGPIVEEDDEINRDWLDWTYSAATFSVFLSILYFYSSLSRFLMVMGATVVMYLHHVGWFPFRPRPVQNFPNDGPPPDVVNQDPNNNLQEGTDPETEDPNHLPPDRDVLDGEQTSPSFMSTAWLVFKTFFASLLPEGPPAIAN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HERP/KIAA0025/Mif1/SUP
Common Name HERPUD1
Gene Symbol HERPUD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.