Learn More
Sino Biological Anti-HEXA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12037T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HEXA (rh HEXA; Catalog#12037-H08B; AAD13932.1; Met1-Thr529). HEXA specific IgG was purified by Human HEXA affinity chromatography.
Catalog No. 50-255-4475