Learn More
Sino Biological Anti-HEXB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200592T40100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human HEXB fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-0960