Learn More
Biomatik Corporation Human HEXB (Beta-hexosaminidase subunit beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58841
Encompass_Preferred
Human HEXB (Beta-hexosaminidase subunit beta) ELISA Kit for detecting HEXB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100iu/ml. Sensitivity: 0.938iu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07686. Target Synonyms: HEXB; ENC-1AS; N-acetyl-beta-glucosaminidase subunit beta; Beta-N-acetylhexosaminidase subunit beta; Hexosaminidase subunit B; Cervical cancer proto-oncogene 7 protein; HCC-7; ENC-1AS; HCC-7; HEL-248; beta-hexosaminidase subunit beta; Beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; hexosaminidase B; beta polypeptide; Hexosaminidase subunit B; N-acetyl-beta-glucosaminidase subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6161