Learn More
Abcam Human Hexokinase 1 ELISA Kit, 1 x 96 Tests
Catalog No. 502698204
Encompass_Preferred
Human Hexokinase 1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Hexokinase 1 with a sensitivity of 301.946 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8204 Supplier Abcam Supplier No. AB3154301X96TESTS
Add to Cart
Add to Cart
Human Hexokinase 1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Hexokinase 1 with a sensitivity of 301.946 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement