Learn More
Biomatik Corporation Human Hexokinase 1 (HK1) ELISA Kit
Supplier:  Biomatik Corporation EKU04772
Encompass_Preferred
Human Hexokinase 1 (HK1) ELISA Kit for detecting HK1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 21.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19367. Gene ID: 3098. Target Synonyms: HK1-Ta; HK1-Tb; HK1-Tc; HKI; HXK1; Brain form hexokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8931