Learn More
Abcam Human HGF ELISA Kit (Hepatocyte Growth Factor), 1 x 96 Tests
Supplier:  Abcam AB2759011X96TESTS
Encompass_Preferred
Human HGF ELISA Kit (Hepatocyte Growth Factor) is a single-wash 90-min Simplestep used to quantify Human HGF (Hepatocyte Growth Factor) with a sensitivity of 18 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2251792