Learn More
Sino Biological HGF / Hepatocyte Growth Factor Neutralizing Antibody
Supplier:  Sino Biological 10463MH010500
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Human HGF / Hepatocyte Growth Factor. The antibody was produced using recombinant antibody technology.
Catalog No. 50-240-9140