Learn More
Sino Biological Anti-HGF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10463RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HGF (rh HGF; Catalog#10463-HNAS; NP_000592.3; Met 1-Ser 728). HGF specific IgG was purified by Human HGF affinity chromatography.
Catalog No. 50-240-9122