Learn More
ACROBiosystems ACROBiosystems Human HGF R-coupled Magnetic Beads
Catalog No. 502107181
Encompass_Preferred
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 61.5 μg S2 protein to 1 mg Beads can bind the Anti-SARS-CoV-2 S2 Antibody with an EC50 of 0.4837 μg/mL (QC tested).
Catalog No. 50-210-7181 Supplier ACROBiosystems Supplier No. MBSK02110MG
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 61.5 μg S2 protein to 1 mg Beads can bind the Anti-SARS-CoV-2 S2 Antibody with an EC50 of 0.4837 μg/mL (QC tested).