Learn More
Gibco™ Human HGF Recombinant Protein, PeproTech®
Description
100-39-1MG will be provided as 4 x 250 μg (100-39-250UG). 100-39-500UG will be provided as 2 x 250 μg (100-39-250UG). Recombinant Human HGF is an 80.0 kDa polypeptide consisting of 697 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P14210 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3082 |
| Molecular Weight (g/mol) | 80 kDa |
| Name | Human HGF |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.