Learn More
Abcam Human HIF-1 alpha ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1715771X96TESTS
Encompass_Preferred
Human HIF-1 alpha ELISA Kit is a single-wash 90-min Simplestep used to quantify Human HIF-1 alpha with a sensitivity of 42 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 15 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2594272