Learn More
Sino Biological Anti-HIF-1 alpha/HIF1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11977RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HIF1A (rh HIF1A; Catalog#11977-H07E; Q16665-1; Arg 575-Asn 826). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1994