Learn More
Biomatik Corporation Human High Density Lipoprotein Binding Protein (HDLBP) ELISA Kit
Supplier:  Biomatik Corporation EKU04783
Encompass_Preferred
Human High Density Lipoprotein Binding Protein (HDLBP) ELISA Kit for detecting HDLBP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00341. Gene ID: 3069. Target Synonyms: HBP; VGL; Vigilin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6235