Learn More
Biomatik Corporation Human High Mobility Group Box Protein 3 (HMGB3) ELISA Kit
Supplier:  Biomatik Corporation EKU10905
Encompass_Preferred
Human High Mobility Group Box Protein 3 (HMGB3) ELISA Kit for detecting HMGB3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15347. Gene ID: 3149. Target Synonyms: HMG2A; HMG4; High mobility group protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5617