Learn More
Biomatik Corporation Human High Temperature Requirement Factor A4 (HTRA4) CLIA Kit
Supplier:  Biomatik Corporation EKU09833
Encompass_Preferred
Human High Temperature Requirement Factor A4 (HTRA4) CLIA Kit for detecting HTRA4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 27.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P83105. Gene ID: 203100. Target Synonyms: HtrA Serine Peptidase 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8422