Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HIP (aa 105-174) Control Fragment Recombinant Protein

Catalog No. RP99511
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99511 Supplier Invitrogen™ Supplier No. RP99511
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83854. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P50502
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6767
Name Human HIP (aa 105-174) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1110007I03Rik; 3110002K08Rik; AAG2; aging-associated protein 2; AW555194; FAM10A1; FAM10A4; heat shock 70kD protein binding protein; hedgehog interacting protein; hedgehog-interacting protein; HHIP; HIP; HOP; Hsc70-interacting protein; Hsp70 interacting protein; Hsp70-interacting protein; HSPABP; HSPABP1; hypothetical protein; p48; PRO0786; Progesterone receptor-associated p48 protein; Protein FAM10A1; Protein ST13 homolog; putative tumor suppressor ST13; RCJMB04_6h13; Renal carcinoma antigen NY-REN-33; SNC6; ST13; ST13 Hsp70 interacting protein; ST13, Hsp70 interacting protein; ST13P5; suppression of tumorigenicity 13; suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein); suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5; suppression of tumorigenicity 13 (colon carcinoma) Hsp70-interacting protein; suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m; UNQ5825/PRO19644
Common Name HIP
Gene Symbol St13
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EITEEMMDQANDKKVAAIEALNDGELQKAIDLFTDAIKLNPRLAILYAKRASVFVKLQKPNAAIRDCDRA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.