Learn More
ACROBiosystems Monoclonal Anti-His Tag Antibody, Human IgG1 (AY63) (MALS verified)
Catalog No. 502719483
Encompass_Preferred
His tag is a very small molecular weight tag, usually composed of 6-10 histidine (His). It is one of the commonly used tags for protein purification and detection. Due to its small molecular weight, fusion into the target protein has almost no effect on the structure and characteristics of the protein. Anti-his tag antibody can accurately detect, locate and purify His tag fusion protein, so as to provide convenience for the vast number of researchers.
Catalog No. 50-271-9483 Supplier ACROBiosystems Supplier No. HISM5351MG
His tag is a very small molecular weight tag, usually composed of 6-10 histidine (His). It is one of the commonly used tags for protein purification and detection. Due to its small molecular weight, fusion into the target protein has almost no effect on the structure and characteristics of the protein. Anti-his tag antibody can accurately detect, locate and purify His tag fusion protein, so as to provide convenience for the vast number of researchers.