Learn More
Sino Biological Anti-H2BC3/HIST1H2BB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101540T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HIST1H2BB, and purified by antigen affinity chromatography.
Catalog No. 50-240-9579