Learn More
Biomatik Corporation Human Histidine Rich Calcium Binding Protein (HRC) ELISA Kit
Supplier:  Biomatik Corporation EKU04804
Encompass_Preferred
Human Histidine Rich Calcium Binding Protein (HRC) ELISA Kit for detecting HRC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23327. Gene ID: 3270. Target Synonyms: HCP; Sarcoplasmic reticulum histidine-rich calcium-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6112