Learn More
Biomatik Corporation Human Histone Cluster 1, H2ab (HIST1H2AB) ELISA Kit
Supplier:  Biomatik Corporation EKU09351
Encompass_Preferred
Human Histone Cluster 1, H2ab (HIST1H2AB) ELISA Kit for detecting HIST1H2AB in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04908. Gene ID: 3012. Target Synonyms: HIST1H2AE; H2AFM; H2A/m; H2A Histone Family Member M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8533