Learn More
Biomatik Corporation Human Histone Cluster 2, H3a (HIST2H3A) ELISA Kit
Supplier:  Biomatik Corporation EKU09361
Encompass_Preferred
Human Histone Cluster 2, H3a (HIST2H3A) ELISA Kit for detecting HIST2H3A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q71DI3. Gene ID: 126961. Target Synonyms: H3/n; H3/o; HIST2H3D; HIST2H3C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7484