Learn More
Biomatik Corporation Human Histone Deacetylase 9 (HDAC9) ELISA Kit
Supplier:  Biomatik Corporation EKU04821
Encompass_Preferred
Human Histone Deacetylase 9 (HDAC9) ELISA Kit for detecting HDAC9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UKV0. Gene ID: 9734. Target Synonyms: HD7; HDAC; HDAC7; HDAC7B; HDAC9B; HDAC9FL; HDRP; MITR; Histone deacetylase-related protein; MEF2-interacting transcription repressor MITR; Histone deacetylase 7B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5474