Learn More
Raybiotech Inc Human HIV-P24 ELISA Kit
Supplier:  Raybiotech Inc ELVHIVP241
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 6.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human HIV-P24.
Catalog No. 50-194-9354