Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HIVEP1 (aa 1523-1667) Control Fragment Recombinant Protein

Catalog No. RP100695
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100695 Supplier Invitrogen™ Supplier No. RP100695
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (50%), Rat (50%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62235. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a transcription factor belonging to the ZAS family, members of which are large proteins that contain a ZAS domain - a modular protein structure consisting of a pair of C2H2 zinc fingers with an acidic-rich region and a serine/threonine-rich sequence. These proteins bind specifically to the DNA sequence motif, GGGACTTTCC, found in the enhancer elements of several viral promoters, including human immunodeficiency virus (HIV), and to related sequences found in the enhancer elements of a number of cellular promoters. This protein binds to this sequence motif, suggesting a role in the transcriptional regulation of both viral and cellular genes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15822
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 3096
Name Human HIVEP1 (aa 1523-1667) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias alpha A-CRYBP1; alpha A-crystallin-binding protein 1; alpha A-crystallin-binding protein I; alphaA-CRYBP1; cirhin interaction protein; CIRIP; Cryabp1; CRYBP1; crystallin, alpha binding protein 1; GAAP; Gate keeper of apoptosis-activating protein; HIV type I enhancer binding protein 1; HIVEP zinc finger 1; HIVEP1; HIV-EP1; human immunodeficiency virus type I enhancer binding protein 1; human immunodeficiency virus type I enhancer-binding protein 1; major histocompatibility complex binding protein 1; Major histocompatibility complex-binding protein 1; MBP-1; positive regulatory domain II binding factor 1; Positive regulatory domain II-binding factor 1; PRDII-BF1; Schnurri-1; si:dkey-184j23.2; si:dkey-31k9.2; transcription factor alphaA-CRYBP1; ZAS1; zinc finger protein 40; ZNF40; ZNF40A
Common Name HIVEP1
Gene Symbol HIVEP1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SHQSTQLSLQVSTQGSKPDKNSVLSGSSKSEDCFAPKYQLHCQVFTSGPSCSSNPVHSLPNQVISDPVGTDHCVTSATLPTKLIDSMSNSHPLLPPELRPLGSQVQKVPSSFMLPIRLQSSVPAYCFATLTSLPQILVTQDLPNQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.