Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human HLA-A*03:01&B2M Monomer Protein, Peptide-Free, C-Terminal His Tag, 25 μg

Catalog No. NC3448933
Encompass_Preferred

Recombinant human HLA-A*03:01&B2M monomer, peptide-free and MALS-verified, intended for immunology research, peptide loading, and binding assays. Produced in HEK293 cells and supplied lyophilized to enable storage and custom reconstitution.

  • Size: 25 μg (also available in 200 μg).
  • Peptide-free, suitable for custom peptide loading.
  • Verified by SEC-MALS, SDS-PAGE, and bioactivity ELISA.
  • Expression system: human 293 (HEK293) cells.
  • Purity: >90% as determined by SDS-PAGE and SEC-MALS.
  • Tag: C-terminal polyhistidine tag for purification or detection.
  • Formulation: lyophilized from PBS (pH 7.4) with trehalose as protectant.
  • Storage: store lyophilized at -20 °C or lower; avoid repeated freeze-thaw after reconstitution.
  • Endotoxin: <1.0 EU/μg (LAL/rFC).
  • Molecular weight: calculated 37.7 kDa (HLA-A*03:01) and 15.7 kDa (B2M); migrates ~42-45 kDa and ~16 kDa on SDS-PAGE.

Catalog No. NC3448933 Supplier ACROBiosystems Supplier No. HLMH52H325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HLA-A*03:01&B2M monomer, peptide-free and MALS-verified, intended for immunology research, peptide loading, and binding assays. Produced in HEK293 cells and supplied lyophilized to enable storage and custom reconstitution.

  • Size: 25 μg (also available in 200 μg).
  • Peptide-free, suitable for custom peptide loading.
  • Verified by SEC-MALS, SDS-PAGE, and bioactivity ELISA.
  • Expression system: human 293 (HEK293) cells.
  • Purity: >90% as determined by SDS-PAGE and SEC-MALS.
  • Tag: C-terminal polyhistidine tag for purification or detection.
  • Formulation: lyophilized from PBS (pH 7.4) with trehalose as protectant.
  • Storage: store lyophilized at -20 °C or lower; avoid repeated freeze-thaw after reconstitution.
  • Endotoxin: <1.0 EU/μg (LAL/rFC).
  • Molecular weight: calculated 37.7 kDa (HLA-A*03:01) and 15.7 kDa (B2M); migrates ~42-45 kDa and ~16 kDa on SDS-PAGE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.