Learn More
ACROBiosystems Unconjugated Human HLA-A*02:01&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719442
Encompass_Preferred
Unconjugated Human HLA-A*02:01&B2M Monomer Protein (HLM-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*02:01) & P61769-1 (B2M)).
Catalog No. 50-271-9442 Supplier ACROBiosystems Supplier No. HLMH52H525UG
Unconjugated Human HLA-A*02:01&B2M Monomer Protein (HLM-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*02:01) & P61769-1 (B2M)).