Learn More
ACROBiosystems Human HLA-A0201 B2M 25ug
Catalog No. 502534702
Encompass_Preferred
PE-Labeled Human HLA-A*0201B2M Tetramer Protein (HLM-HP2H3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M)).
Catalog No. 50-253-4702 Supplier ACROBiosystems Supplier No. HLMHP2H325UG
PE-Labeled Human HLA-A*0201B2M Tetramer Protein (HLM-HP2H3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M)).