Learn More
ACROBiosystems Human HLA-A0201B2MPRAME S
Catalog No. 502534732
Encompass_Preferred
Human HLA-A*0201B2MPRAME (SLLQHLIGL) Tetramer Protein (HLE-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (B2M) Gly 25 - Ile 308 (HLA-A*0201) SLLQHLIGL peptide (Accession # P61769 (B2M) AAA59606.1 (HLA-A*0201) SLLQHLIGL).
Catalog No. 50-253-4732 Supplier ACROBiosystems Supplier No. HLEH52H925UG
Human HLA-A*0201B2MPRAME (SLLQHLIGL) Tetramer Protein (HLE-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (B2M) Gly 25 - Ile 308 (HLA-A*0201) SLLQHLIGL peptide (Accession # P61769 (B2M) AAA59606.1 (HLA-A*0201) SLLQHLIGL).