Learn More
ACROBiosystems PE-Labeled Human HLA-A*03:01&B2M Tetramer Protein (Peptide free)
Catalog No. 502719419
Encompass_Preferred
Major histocompatibility complex (MHC), group of genes that code for proteins found on the surfaces of cells that help the immune system recognize foreign substances. MHC proteins are found in all higher vertebrates. In human beings the complex is also called the human leukocyte antigen (HLA) system.
Catalog No. 50-271-9419 Supplier ACROBiosystems Supplier No. HLMHP2H6200UG
Major histocompatibility complex (MHC), group of genes that code for proteins found on the surfaces of cells that help the immune system recognize foreign substances. MHC proteins are found in all higher vertebrates. In human beings the complex is also called the human leukocyte antigen (HLA) system.