Learn More
ACROBiosystems PE-Labeled Human HLA-A*11:01&B2M Tetramer Protein (Peptide free)
Catalog No. 502719421
Encompass_Preferred
Human papillomavirus (HPV) is A kind of papillomavirus belonging to the milk polypoid virus family. It is a spherical DNA virus, which can cause the proliferation of squamous epithelium of human skin mucosa. HPV(human papillomavirus) for common warts, genital warts (condyloma acuminatum), and other symptoms. There are many types of human papillomavirus (HPV), with HPV 16 and 18 being high-risk types known to significantly increase the risk of cervical, vaginal and vulvar cancers in women and men. The PE-labeled human HLA-A*1101 HPV (IVCPICSQK) tetramer protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and IVCPICSQK peptide of the HPV.
Catalog No. 50-271-9421 Supplier ACROBiosystems Supplier No. HLMHP2H5200UG
Human papillomavirus (HPV) is A kind of papillomavirus belonging to the milk polypoid virus family. It is a spherical DNA virus, which can cause the proliferation of squamous epithelium of human skin mucosa. HPV(human papillomavirus) for common warts, genital warts (condyloma acuminatum), and other symptoms. There are many types of human papillomavirus (HPV), with HPV 16 and 18 being high-risk types known to significantly increase the risk of cervical, vaginal and vulvar cancers in women and men. The PE-labeled human HLA-A*1101 HPV (IVCPICSQK) tetramer protein is a complex of HLA-A*1101 of the MHC Class I, B2M, and IVCPICSQK peptide of the HPV.