Learn More
ACROBiosystems Unconjugated Human HLA-A*24:02&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719444
Encompass_Preferred
Unconjugated Human HLA-A*24:02&B2M Monomer Protein (HLM-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M)).
Catalog No. 50-271-9444 Supplier ACROBiosystems Supplier No. HLMH52E425UG
Unconjugated Human HLA-A*24:02&B2M Monomer Protein (HLM-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M)).