Learn More
ACROBiosystems PE-Labeled Human HLA-A*24:02&B2M Tetramer Protein (Peptide free)
Catalog No. 502719422
Encompass_Preferred
PE-Labeled Human HLA-A*24:02&B2M Tetramer Protein (HLM-HP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M) ).
Catalog No. 50-271-9422 Supplier ACROBiosystems Supplier No. HLMHP2H425UG
PE-Labeled Human HLA-A*24:02&B2M Tetramer Protein (HLM-HP2H4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*24:02) & Ile 21 - Met 119 (B2M) (Accession # AAA59600.1 (HLA-A*24:02) & P61769 (B2M) ).