Learn More
ACROBiosystems Biotinylated Human HLA-A*26:01&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719429
Encompass_Preferred
Biotinylated Human HLA-A*26:01&B2M Monomer Protein (HLM-H82Ev) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Trp 298 (HLA-A*26:01) & Ile 21 - Met 119 (B2M) (Accession # AOE46674.1 (HLA-A*26:01) & P61769 (B2M)).
Catalog No. 50-271-9429 Supplier ACROBiosystems Supplier No. HLMH82EV200UG
Biotinylated Human HLA-A*26:01&B2M Monomer Protein (HLM-H82Ev) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Trp 298 (HLA-A*26:01) & Ile 21 - Met 119 (B2M) (Accession # AOE46674.1 (HLA-A*26:01) & P61769 (B2M)).