Learn More
ACROBiosystems Human HLA-B0702 B2M 200ug
Catalog No. 502534713
Encompass_Preferred
Biotinylated Human HLA-B*0702B2M Monomer Protein (HLM-H82Ek) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*0702) Ile 21 - Met 119 (B2M) (Accession # QDK63242.1 (HLA-B*0702) P61769 (B2M)).
Catalog No. 50-253-4713 Supplier ACROBiosystems Supplier No. HLMH82EK200UG
Biotinylated Human HLA-B*0702B2M Monomer Protein (HLM-H82Ek) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*0702) Ile 21 - Met 119 (B2M) (Accession # QDK63242.1 (HLA-B*0702) P61769 (B2M)).