Learn More
ACROBiosystems Human HLA-B0801 B2M 25ug
Catalog No. 502534708
Encompass_Preferred
Biotinylated Human HLA-B*0801B2M Monomer Protein (HLM-H82En) is expressed from human 293 cells (HEK293). It contains AA Gly 25-Val 309 (HLA-B*0801) Ile 21-Met 119 (B2M) (Accession # BAA22205.1 (HLA-B*0801) P61769 (B2M)).
Catalog No. 50-253-4708 Supplier ACROBiosystems Supplier No. HLMH82EN25UG
Biotinylated Human HLA-B*0801B2M Monomer Protein (HLM-H82En) is expressed from human 293 cells (HEK293). It contains AA Gly 25-Val 309 (HLA-B*0801) Ile 21-Met 119 (B2M) (Accession # BAA22205.1 (HLA-B*0801) P61769 (B2M)).