Learn More
ACROBiosystems Human HLA-B1501 B2M 200ug
Catalog No. 502534705
Encompass_Preferred
Biotinylated Human HLA-B*1501B2M Monomer Protein (HLM-H82W8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*1501) Ile 21 - Met 119 (B2M) (Accession # SCQ83648.1 (HLA-B*1501) P61769 (B2M)).
Catalog No. 50-253-4705 Supplier ACROBiosystems Supplier No. HLMH82W8200UG
Biotinylated Human HLA-B*1501B2M Monomer Protein (HLM-H82W8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*1501) Ile 21 - Met 119 (B2M) (Accession # SCQ83648.1 (HLA-B*1501) P61769 (B2M)).