Learn More
ACROBiosystems Biotinylated Human HLA-B*40:01&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719431
Encompass_Preferred
Biotinylated Human HLA-B*40:01&B2M Monomer Protein (HLM-H82Eu) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*40:01) & Ile 21 - Met 119 (B2M) (Accession # AAA03719.1 (HLA-B*40:01) & P61769 (B2M)).
Catalog No. 50-271-9431 Supplier ACROBiosystems Supplier No. HLMH82EU200UG
Biotinylated Human HLA-B*40:01&B2M Monomer Protein (HLM-H82Eu) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*40:01) & Ile 21 - Met 119 (B2M) (Accession # AAA03719.1 (HLA-B*40:01) & P61769 (B2M)).