Learn More
ACROBiosystems Biotinylated Human HLA-C*03:03&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719440
Encompass_Preferred
Biotinylated Human HLA-C*03:03&B2M Monomer Protein (HLM-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 25- Ile 308 (HLA-C*03:03) & Ile 21-Met 119 (B2M) (Accession # QDK63961.1 (HLA-C*03:03) & P61769-1 (B2M)).
Catalog No. 50-271-9440 Supplier ACROBiosystems Supplier No. HLMH82E925UG
Biotinylated Human HLA-C*03:03&B2M Monomer Protein (HLM-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 25- Ile 308 (HLA-C*03:03) & Ile 21-Met 119 (B2M) (Accession # QDK63961.1 (HLA-C*03:03) & P61769-1 (B2M)).