Learn More
ACROBiosystems Biotinylated Human HLA-C*03:04&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719437
Encompass_Preferred
Biotinylated Human HLA-C*03:04&B2M Monomer Protein (HLM-H82Er) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-C*03:04) & Ile 21 - Met 119 (B2M) (Accession # CAH6418125.1 (HLA-C*03:04) & P61769 (B2M)).
Catalog No. 50-271-9437 Supplier ACROBiosystems Supplier No. HLMH82ER200UG
Biotinylated Human HLA-C*03:04&B2M Monomer Protein (HLM-H82Er) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-C*03:04) & Ile 21 - Met 119 (B2M) (Accession # CAH6418125.1 (HLA-C*03:04) & P61769 (B2M)).